missing translation for 'onlineSavingsMsg'
Learn More

2-(4-tert-butoxycarbonylpiperazin-1-yl)pyridine-4-boronic acid, 97%, Thermo Scientific™

Product Code. 10072124
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10072124 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10072124 Supplier Thermo Scientific Maybridge Supplier No. MAY00032DA
 Restricted Item
Estimated Shipment: 02-09-2026
Add to basket
This item is not returnable. View return policy

Specifications

CAS 936250-21-4
CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Molecular Formula C20H32BN3O4
MDL Number MFCD06797996
Quantity 1 g
Synonym 2-4-tert-butoxycarbonylpiperazin-1-yl pyridine-4-boronic acid, pinacol ester, tert-butyl 4-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridin-2-yl piperazine-1-carboxylate, 2-4-boc-piperazino pyridine-4-boronic acid, pinacol ester, 2-4-tert-butoxycarbonyl piperazin-1-yl-4-4,4,5,5-tetramethyl,1,3,2-dioxaborolan-2-yl pyridine, 2-4-boc-piperazino pyridine-4-boronic acid pinacol ester, 2-4-boc-piperazin-1-yl pyridine-4-boronic acid pinacol ester, 2-4-n-boc-piperazin-1-yl pyridine-4-boronic acid, pinacol ester, 2-4-tert-butoxycarbonylpiperazinyl pyridine-4-boronic acid, pinacol ester, tert-butyl 4-4-tetramethyl-1,3,2-dioxaborolan-2-yl pyridin-2-yl piperazine-1-carboxylate
InChI Key NQBNJXYYAKBREJ-UHFFFAOYSA-N
SMILES CC(C)(C)OC(=O)N1CCN(CC1)C1=NC=CC(=C1)B1OC(C)(C)C(C)(C)O1
IUPAC Name tert-butyl 4-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-2-yl]piperazine-1-carboxylate
Molecular Weight (g/mol) 389.30
PubChem CID 17998916
Formula Weight 389.3
Percent Purity 97%
Chemical Name or Material 2-(4-tert-Butoxycarbonylpiperazin-1-yl)pyridine-4-boronic acid, pinacol ester
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.