missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals 2-(4-Sulfophenylazo)1,8-dihydroxy-3,6-naphthalenedisulfonic Acid, Trisodium Salt, Indicator Grade

Product Code. 10064623 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
10 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
10g
25g
This item is not returnable. View return policy

Product Code. 10064623

Brand: Thermo Scientific Chemicals 151571000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 23647-14-5 | C16H9N2Na3O11S3 | 570.39 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 23647-14-5
Molecular Formula C16H9N2Na3O11S3
Molecular Weight (g/mol) 570.39
MDL Number MFCD00003952
InChI Key LSAWBVPBZOEGOT-UHFFFAOYSA-L
Synonym spadns, 2,7-naphthalenedisulfonic acid, 4,5-dihydroxy-3-4-sulfophenyl azo-, trisodium salt, trisodium 4,5-dihydroxy-3-e-2-4-sulfonatophenyl diazen-1-yl naphthalene-2,7-disulfonate, 2,7-naphthalenedisulfonic acid, 4,5-dihydroxy-3-2-4-sulfophenyl diazenyl-, sodium salt 1:3, 2-p-sulfophenylazo-1,8-dihydroxy-3,6-naph-thalenedisulfonic acid
PubChem CID 73658865
IUPAC Name trisodium;5-hydroxy-4-oxo-3-[(4-sulfophenyl)hydrazinylidene]naphthalene-2,7-disulfonate
SMILES C1=CC(=CC=C1NN=C2C(=CC3=CC(=CC(=C3C2=O)O)S(=O)(=O)[O-])S(=O)(=O)[O-])S(=O)(=O)O.[Na+].[Na+].[Na+]

Specifications

Color Brown-Red
Infrared Spectrum Authentic
Packaging Glass bottle
Quantity 100 g
Merck Index 15, 8859
Solubility Information Solubility in water: soluble. Other solubilities: practically insoluble in organic solvents
Formula Weight 570.39
Grade Indicator
Physical Form Crystalline Powder
Chemical Name or Material 2-(4-Sulfophenylazo)1,8-dihydroxy-3,6-naphthalenedisulfonic acid, trisodium salt

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.