missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-[4-[(RS)-(4-Chlorophenyl)phenylmethyl]piperazin-1-yl]ethanol Dihydrochloride, Mikromol™
Description
2-[4-[(RS)-(4-Chlorophenyl)phenylmethyl]piperazin-1-yl]ethanol Dihydrochloride
Specifications
Specifications
| Chemical Name or Material | 4-[(4-Chlorophenyl)phenylmethyl]-1-piperazineethanol Dihydrochloride |
| CAS | 164726-80-1 |
| Analyte or Component Names | 4-[(4-Chlorophenyl)phenylmethyl]-1-piperazineethanol Dihydrochloride |
| Shipping Condition | Room Temperature |
| Molecular Formula | C19 H23 Cl N2 O . 2 Cl H |
| Certifications/Compliance | ISO 17025 |
| Synonym | Cetirizine Dihydrochloride Imp. G (EP), Cetirizine Imp. G (EP), Cetirizine USP Related Compound G, Cetirizine USP RC G, 2-[4-[(RS)-(4-Chlorophenyl)phenylmethyl]piperazin-1-yl]ethanol Dihydrochloride, Cetirizine Dihydrochloride Imp. G (EP) as Dihydrochloride, 4-[(4-Chlorophenyl)phenylmethyl]-1-piperazine-ethanol Dihydrochloride, 1-Piperazineethanol, 4-[(4-chlorophenyl)phenylmethyl]-, hydrochloride (1:2), 1-Piperazineethanol, 4-[(4-chlorophenyl)phenylmethyl]-, dihydrochloride (9CI), Cetirizine Related Compound G as Dihydrochloride, Cetirizine Dihydrochloride Impurity G as Dihydrochloride, Cetirizine Impurity G as Dihydrochloride |
| InChI Formula | InChI=1S/C19H23ClN2O.2ClH/c20-18-8-6-17(7-9-18)19(16-4-2-1-3-5-16)22-12-10-21(11-13-22)14-15-23;;/h1-9,19,23H,10-15H2;2*1H |
| SMILES | Cl.Cl.OCCN1CCN(CC1)C(c2ccccc2)c3ccc(Cl)cc3 |
| IUPAC Name | 2-[4-[(4-chlorophenyl)-phenylmethyl]piperazin-1-yl]ethanol;dihydrochloride |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?