missing translation for 'onlineSavingsMsg'
Learn More

2-(4-Pyridyl)malondialdehyde, 95%

Product Code. 10536992 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
2.5 g
Packaging:
Glass bottle
Unit Size:
2.5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10536992 2.5 g Glass bottle 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10536992 Supplier Thermo Scientific Acros Supplier No. 425340025
 Restricted Item
Estimated Shipment: 15-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 51076-46-1 | C8H7NO2 | 149.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 51076-46-1
CAS Min % 94.0
CAS Max % 100.0
Melting Point >320.0°C
Color Beige to Brown
Infrared Spectrum Authentic
Assay Percent Range 94% min
Packaging Glass bottle
Molecular Formula C8H7NO2
MDL Number MFCD00216535
Quantity 2.5 g
Synonym 2-4-pyridyl malondialdehyde, 2-pyridin-4-yl malonaldehyde, 2-pyridin-4-yl propanedial, propanedial, 4-pyridinyl, 2-4-pyridinyl malonaldehyde, 2-pyridin-4-yl malondialdehyde, zlchem 842, 2-4-pyridyl propanedial, ksc925a8h, 2-4-pyridyl propane-1,3-dial
Solubility Information Solubility in water: slightly soluble.
InChI Key RYYVVCNGQOENKM-UHFFFAOYSA-N
SMILES C1=CN=CC=C1C(C=O)C=O
IUPAC Name 2-pyridin-4-ylpropanedial
Molecular Weight (g/mol) 149.15
PubChem CID 2737246
Formula Weight 149.15
Percent Purity 95%
Physical Form Powder
Chemical Name or Material 2-(4-Pyridyl)malondialdehyde, 95%
Show More Show Less
compliance-icons
Product Identifier
  • 2-(4-Pyridyl)malondialdehyde
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.