missing translation for 'onlineSavingsMsg'
Learn More

2,4-Pyridinedicarboxylic acid hydrate, 99+%

Product Code. 10376870 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10376870 5 g Glass bottle 5g
10720595 1 g Glass bottle 1g
10540994 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10376870 Supplier Thermo Scientific Acros Supplier No. 131860050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 499-80-9 | C7H5NO4 | 167.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 499-80-9
Molecular Formula C7H5NO4
Molecular Weight (g/mol) 167.12
MDL Number MFCD00149413
InChI Key MJIVRKPEXXHNJT-UHFFFAOYSA-N
Synonym 2,4-pyridinedicarboxylic acid, lutidinic acid, 2,4-lutidinic acid, 2,4-pyridinedicarboxylicacid, hydrate 1:1, 4nrq, pd2, 4-carboxypicolinic acid, pubchem16314, pyridine carboxylate, 6a, acmc-1bnb9
PubChem CID 10365
ChEBI CHEBI:44737
IUPAC Name pyridine-2,4-dicarboxylic acid
SMILES OC(=O)C1=CC=NC(=C1)C(O)=O

Specifications

CAS Min % 99.0
CAS Max % 100.0
Melting Point 243°C to 246°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Glass bottle
Linear Formula (C5H3N)(COOH)2·xH2O
Quantity 5 g
Beilstein 22, 153
Solubility Information Solubility in water: insoluble
Formula Weight 167.12
Percent Purity 99+%
Physical Form Crystalline Powder
Chemical Name or Material 2, 4-Pyridinedicarboxylic acid hydrate, 99+%
Show More Show Less
Product Identifier
  • 2,4-Pyridinedicarboxylic acid hydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.