missing translation for 'onlineSavingsMsg'
Learn More

2,4-Pentadienoic acid, 97%, stabilized

Product Code. p-7077576 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15375038 1 g 1g
15395038 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15375038 Supplier Thermo Scientific Acros Supplier No. 455740010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 626-99-3 | C5H6O2 | 98.1 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 626-99-3
Molecular Formula C5H6O2
Molecular Weight (g/mol) 98.1
InChI Key SDVVLIIVFBKBMG-ONEGZZNKSA-N
Synonym penta-2,4-dienoic acid, 2,4-pentadienoic acid, trans-2,4-pentadienoic acid, 1,3-butadiene-1-carboxylic acid, e-2,4-pentadienoic acid, e-penta-2,4-dienoic acid, 2e-2,4-pentadienoic acid, beta-vinyl acrylic acid, vynilacrylic acid, 2e-penta-2,4-dienoic acid
PubChem CID 642034
ChEBI CHEBI:37331
IUPAC Name (2E)-penta-2,4-dienoic acid
SMILES C=CC=CC(=O)O

Specifications

Melting Point 69.0°C to 72.0°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass Bottle
Linear Formula CH2(CH)3COOH
Quantity 1 g
Solubility Information Solubility in water: soluble.
Formula Weight 98.1
Percent Purity 97%
Chemical Name or Material 2,4-Pentadienoic acid
compliance-icons
Product Identifier
  • 2,4-Pentadienoic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P362-Take off contaminated clothing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.