missing translation for 'onlineSavingsMsg'
Learn More

2-(4-Nitrophenyl)ethylamine hydrochloride, 98+%

Product Code. p-7056133
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15412299 1 g 1g
15422299 5 g 5g
15432299 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15412299 Supplier Thermo Scientific Alfa Aesar Supplier No. H64894.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 29968-78-3 | C8H11N2O2 | 167.19 g/mol

4-Nitrophenethylamine hydrochloride was used in the synthesis of ortho-metalated primary phenethylamines complexes containing six-membered palladacycles and N-(4-nitrophenethyl)formamide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Nitrophenethylamine hydrochloride was used in the synthesis of ortho-metalated primary phenethylamines complexes containing six-membered palladacycles and N-(4-nitrophenethyl)formamide.

Solubility
Soluble in methanol almost transparency.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 29968-78-3
Molecular Formula C8H11N2O2
Molecular Weight (g/mol) 167.19
MDL Number MFCD00012900
InChI Key IOXOZOPLBFXYLM-UHFFFAOYSA-O
Synonym 4-nitrophenethylamine hydrochloride, 4-nitrophenylethylamine hydrochloride, 4-nitrophenethylamine hcl, 2-4-nitrophenyl ethanamine hydrochloride, 2-4-nitrophenyl ethylamine hydrochloride, p-nitrophenylethylamine hcl, benzeneethanamine, 4-nitro-, monohydrochloride, 4-nitrophenethylaminehydrochloride, p-nitrophenethylamine, hydrochloride, pubchem11627
PubChem CID 458983
IUPAC Name 2-(4-nitrophenyl)ethanamine;hydrochloride
SMILES [NH3+]CCC1=CC=C(C=C1)[N+]([O-])=O

Specifications

Melting Point 200°C (literature)
Quantity 1 g
Beilstein 3568915
Solubility Information Soluble in methanol almost transparency.
Formula Weight 202.64
Percent Purity ≥98%
Physical Form Crystalline Powder
Chemical Name or Material 2-(4-Nitrophenyl)ethylamine hydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.