missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-(4-n-Propylphenyl)propanoic Acid, Mikromol™
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
Description
2-(4-n-Propylphenyl)propanoic Acid
Specifications
Specifications
| Chemical Name or Material | 2-(4-n-Propylphenyl)propanoic Acid |
| CAS | 3585-47-5 |
| Analyte or Component Names | 2-(4-n-Propylphenyl)propanoic Acid |
| Shipping Condition | Ice pack (-20°C) |
| Molecular Formula | C12 H16 O2 |
| Certifications/Compliance | ISO 17025 |
| Synonym | 2-(4-n-Propylphenyl)propanoic Acid |
| InChI Formula | InChI=1S/C12H16O2/c1-3-4-10-5-7-11(8-6-10)9(2)12(13)14/h5-9H,3-4H2,1-2H3,(H,13,14) |
| SMILES | CCCc1ccc(cc1)C(C)C(=O)O |
| IUPAC Name | 2-(4-propylphenyl)propanoic acid |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto