missing translation for 'onlineSavingsMsg'
Learn More

2-(4-Morpholinyl)pyridine, 96%, Thermo Scientific™

Product Code. 15452398
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 24255-25-2
Molecular Formula C9H12N2O
Molecular Weight (g/mol) 164.21
MDL Number MFCD01143321
InChI Key XHPVOSNOIWGRQQ-UHFFFAOYSA-N
Synonym 4-pyridin-2-yl morpholine, 2-morpholinopyridine, morpholinopyridine, 4-2-pyridinyl morpholine, 4-pyridin-2-yl-morpholine, 2-4-morpholinyl pyridine, 2-morpholinopyridine 96
PubChem CID 10964853
IUPAC Name 4-pyridin-2-ylmorpholine
SMILES C1CN(CCO1)C1=CC=CC=N1

Specifications

Density 1.131 g/mL
Boiling Point 135°C (20 mmHg)
Flash Point >110°C (230°F)
Quantity 1 g
Formula Weight 164.21
Percent Purity 96%
Physical Form Liquid
Chemical Name or Material 2-(4-Morpholinyl)pyridine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'