Learn More
[2-(4-Methylpiperazin-1-yl)phenyl]methanol, 97%, Thermo Scientific™
£18.40 - £324.00
Chemical Identifiers
CAS | 123987-12-2 |
---|---|
Molecular Formula | C12H19N2O |
Molecular Weight (g/mol) | 207.30 |
MDL Number | MFCD03407383 |
InChI Key | TWPYBKBPHCMUIS-UHFFFAOYSA-O |
Synonym | 2-4-methylpiperazin-1-yl phenyl methanol, 2-4-methylpiperazin-1-yl benzyl alcohol, 2-4-methylpiperazino benzyl alcohol, 2-4-methylpiperazin-1yl phenyl methanol, pubchem12999, 2-4-methylpiperazin benzyl alcohol, 2-4-n-methylpiperazinyl benzyl alcohol, 2-4-methyl-1-piperazinyl phenyl methanol, 2-4-methylpiperazinyl phenyl methan-1-ol |
PubChem CID | 2795567 |
SMILES | C[NH+]1CCN(CC1)C1=CC=CC=C1CO |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10619184
|
Thermo Scientific Maybridge
CC36009DA |
1 mg |
£39.15
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10567863
|
Thermo Scientific Maybridge
CC36009CB |
250 mg |
£18.40
250mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10337354
|
Thermo Scientific Maybridge
CC36009EA |
10 g |
£324.00
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Chemical Identifiers
123987-12-2 | |
207.30 | |
TWPYBKBPHCMUIS-UHFFFAOYSA-O | |
2795567 |
C12H19N2O | |
MFCD03407383 | |
2-4-methylpiperazin-1-yl phenyl methanol, 2-4-methylpiperazin-1-yl benzyl alcohol, 2-4-methylpiperazino benzyl alcohol, 2-4-methylpiperazin-1yl phenyl methanol, pubchem12999, 2-4-methylpiperazin benzyl alcohol, 2-4-n-methylpiperazinyl benzyl alcohol, 2-4-methyl-1-piperazinyl phenyl methanol, 2-4-methylpiperazinyl phenyl methan-1-ol | |
C[NH+]1CCN(CC1)C1=CC=CC=C1CO |
Specifications
123987-12-2 | |
100.0 | |
Amber glass bottle | |
MFCD03407383 | |
TWPYBKBPHCMUIS-UHFFFAOYSA-O | |
[2-(4-methylpiperazin-1-yl)phenyl]methanol | |
2795567 | |
97% |
96.5 | |
97% | |
C12H19N2O | |
2-4-methylpiperazin-1-yl phenyl methanol, 2-4-methylpiperazin-1-yl benzyl alcohol, 2-4-methylpiperazino benzyl alcohol, 2-4-methylpiperazin-1yl phenyl methanol, pubchem12999, 2-4-methylpiperazin benzyl alcohol, 2-4-n-methylpiperazinyl benzyl alcohol, 2-4-methyl-1-piperazinyl phenyl methanol, 2-4-methylpiperazinyl phenyl methan-1-ol | |
C[NH+]1CCN(CC1)C1=CC=CC=C1CO | |
207.30 | |
206.29 | |
[2-(4-Methylpiperazin-1-yl)phenyl]methanol |
Safety and Handling
GHS H Statement:
Harmful if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes serious eye damage.
Causes severe skin burns and eye damage.
GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specifica
GHS Signal Word: Danger