missing translation for 'onlineSavingsMsg'
Learn More

{2-[(4-Methylpiperazin-1-yl)methyl]phenyl}methylamine, 97%, Thermo Scientific™

Product Code. 10042303
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 10042303

Brand: Thermo Scientific Maybridge CC47413DA

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 879896-50-1
Molecular Formula C13H21N3
Molecular Weight (g/mol) 219.332
MDL Number MFCD06797802
InChI Key OKGWICVOLZFMPE-UHFFFAOYSA-N
Synonym 2-4-methylpiperazin-1-ylmethyl benzylamine, 1-2-4-methylpiperazin-1-yl methyl phenyl methanamine, 2-4-methylpiperazin-1-yl methyl phenyl methanamine, 2-4-methylpiperazinyl methyl phenyl methylamine, 2-4-methyl-1-piperazinyl methyl-benzenemethanamine, 2-4-methylpiperazin-1-yl methyl phenyl methylamine, benzenemethanamine,2-4-methyl-1-piperazinyl methyl
PubChem CID 16228064
IUPAC Name [2-[(4-methylpiperazin-1-yl)methyl]phenyl]methanamine
SMILES CN1CCN(CC1)CC2=CC=CC=C2CN

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 219.33
Percent Purity 97%
Chemical Name or Material {2-[(4-Methylpiperazin-1-yl)methyl]phenyl}methylamine
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.