missing translation for 'onlineSavingsMsg'
Learn More

2-(4-Methylphenyl)thiazole-4-carboxylic acid, 97%, Thermo Scientific™

Product Code. 15420706
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15420706 250 mg 250mg
15460478 1 g 1g
15470478 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15420706 Supplier Thermo Scientific Alfa Aesar Supplier No. H54054.MD
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Used as pharmaceutical intermediates.

Solubility
Slightly soluble in water.

Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17228-99-8
Molecular Formula C11H8NO2S
Molecular Weight (g/mol) 218.25
MDL Number MFCD01936002
InChI Key MTVWIZYQYPRZGZ-UHFFFAOYSA-M
PubChem CID 2794795
IUPAC Name 2-(4-methylphenyl)-1,3-thiazole-4-carboxylic acid
SMILES CC1=CC=C(C=C1)C1=NC(=CS1)C([O-])=O

Specifications

Melting Point 168°C to 172°C
Quantity 250 mg
Synonym 2-4-methylphenyl-1,3-thiazole-4-carboxylic acid, 2-p-tolyl thiazole-4-carboxylic acid, 2-4-methylphenyl thiazole-4-carboxylic acid, 2-p-tolylthiazole-4-carboxylic acid, 2-p-tolyl-thiazole-4-carboxylic acid, 4-thiazolecarboxylicacid,2-4-methylphenyl, 4-thiazolecarboxylicacid, 2-4-methylphenyl, 2-4-methylphenyl-1,3-thiazole-4-carboxylicacid
Solubility Information Slightly soluble in water.
Formula Weight 219.26
Percent Purity 97%
Chemical Name or Material 2-(4-Methylphenyl)thiazole-4-carboxylic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.