missing translation for 'onlineSavingsMsg'
Learn More

2-(4-Methylphenyl)-1,3-thiazole-4-carboxylic acid, 97%, Thermo Scientific™

Product Code. 10183314
Change view
Click to view available options
Quantity:
250 mg
1 g
10 g
Unit Size:
10g
1g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10183314 10 g 10g
10463943 250 mg 250mg
10003952 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10183314 Supplier Thermo Scientific Maybridge Supplier No. MO07123EA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 17228-99-8
Molecular Formula C11H8NO2S
Molecular Weight (g/mol) 218.25
MDL Number MFCD01936002
InChI Key MTVWIZYQYPRZGZ-UHFFFAOYSA-M
Synonym 2-4-methylphenyl-1,3-thiazole-4-carboxylic acid, 2-p-tolyl thiazole-4-carboxylic acid, 2-4-methylphenyl thiazole-4-carboxylic acid, 2-p-tolylthiazole-4-carboxylic acid, 2-p-tolyl-thiazole-4-carboxylic acid, 4-thiazolecarboxylicacid,2-4-methylphenyl, 4-thiazolecarboxylicacid, 2-4-methylphenyl, 2-4-methylphenyl-1,3-thiazole-4-carboxylicacid
PubChem CID 2794795
SMILES CC1=CC=C(C=C1)C1=NC(=CS1)C([O-])=O

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Quantity 10 g
IUPAC Name 2-(4-methylphenyl)-1,3-thiazole-4-carboxylic acid
Formula Weight 219.26
Percent Purity 97%
Chemical Name or Material 2-(4-Methylphenyl)-1, 3-thiazole-4-carboxylic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.