missing translation for 'onlineSavingsMsg'
Learn More

[2-(4-Methylperhydro-1,4-diazepin-1-yl)phenyl]methanol, ≥97%, Thermo Scientific™

Product Code. 11336845
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11336845

Brand: Thermo Scientific Maybridge CC60409DE

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 915707-55-0
Molecular Formula C13H20N2O
Molecular Weight (g/mol) 220.32
MDL Number MFCD09065000
InChI Key IYBDLMFCIKDQDV-UHFFFAOYSA-N
Synonym 2-4-methylperhydro-1,4-diazepin-1-yl phenyl methanol, 2-4-methyl-1,4-diazepan-1-yl phenyl methanol, 2-4-methylhomopiperazin-1-yl benzyl alcohol, 2-4-methyl-1,4-diazaperhydroepinyl phenyl methan-1-ol
PubChem CID 24229618
IUPAC Name [2-(4-methyl-1,4-diazepan-1-yl)phenyl]methanol
SMILES CN1CCCN(CC1)C1=CC=CC=C1CO

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97+%
Packaging Amber Glass Bottle
Quantity 5 g
Formula Weight 220.31
Percent Purity 97%
Chemical Name or Material [2-(4-Methylperhydro-1, 4-diazepin-1-yl)phenyl]methanol
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.