missing translation for 'onlineSavingsMsg'
Learn More

2-(4-Methylperhydro-1,4-diazepin-1-yl)benzoic acid hydrochloride hemihydrate, 95%, Thermo Scientific™

Product Code. 10103774
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10103774 5 g Amber Glass Bottle 5g
10602895 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10103774 Supplier Thermo Scientific Maybridge Supplier No. CC60451DE
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 921938-77-4
Molecular Formula C13H19ClN2O2
Molecular Weight (g/mol) 270.757
MDL Number MFCD09065004
InChI Key UCYNEWGETFFMQY-UHFFFAOYSA-N
Synonym 2-4-methyl-1,4-diazepan-1-yl benzoic acid hydrochloride, 2-4-methyl-1,4-diazepan-1-yl benzoic acid hcl, 2-4-methylhomopiperazin-1-yl benzoic acid hydrochloride hemihydrate, 2-4-methyl-1,4-diazepan-1-yl benzoic acid-hydrogen chloride 1/1, 2-4-methylperhydro-1,4-diazepin-1-yl benzoic acid hydrochloride 0.5 hydrate, benzoicacid, 2-hexahydro-4-methyl-1h-1,4-diazepin-1-yl-, hydrochloride 1:1, benzoicacid,2-hexahydro-4-methyl-1h-1,4-diazepin-1-yl-,hydrochloride 1:1
PubChem CID 24229621
IUPAC Name 2-(4-methyl-1,4-diazepan-1-yl)benzoic acid;hydrochloride
SMILES CN1CCCN(CC1)C2=CC=CC=C2C(=O)O.Cl

Specifications

CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 5 g
Formula Weight 279.77
Percent Purity 95%
Chemical Name or Material 2-(4-Methylperhydro-1, 4-diazepin-1-yl)benzoic acid hydrochloride hemihydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.