missing translation for 'onlineSavingsMsg'
Learn More

2-(((4-Methyl-7-oxyooctyl)oxy)carbonyl)cyclohexanecarboxylic Acid (Mixture of Diastereomers), TRC

Product Code. 30006316
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30006316 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30006316 Supplier Toronto Research Chemicals Supplier No. TRCM3292001MG
 Restricted Item
Estimated Shipment: 02-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 1588520-62-0
Molecular Formula C17H28O5
Quantity 1 mg
Synonym Oxo-MINCH
InChI Formula InChI=1S/C17H28O5/c1-12(9-10-13(2)18)6-5-11-22-17(21)15-8-4-3-7-14(15)16(19)20/h12,14-15H,3-11H2,1-2H3,(H,19,20)
SMILES CC(CCCOC(=O)C1CCCCC1C(=O)O)CCC(=O)C
IUPAC Name 2-(4-methyl-7-oxo-octoxy)carbonylcyclohexanecarboxylic acid
Molecular Weight (g/mol) 312.4
Formula Weight 312.1937
Percent Purity >95%
Chemical Name or Material Cyclohexane-1,2-dicarboxylic acid, mono-(4-methyl-7-oxo octyl)ester
Grade HPLC
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.