missing translation for 'onlineSavingsMsg'
Learn More

2-(4-Methyl-1,3-thiazol-2-yl)acetonitrile, 97%, Thermo Scientific™

Product Code. 10143424
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10143424 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10143424 Supplier Thermo Scientific Maybridge Supplier No. SB02109EA
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 19785-39-8
Molecular Formula C6H6N2S
Molecular Weight (g/mol) 138.19
MDL Number MFCD03011392
InChI Key XSRRCOBFMZWKJR-UHFFFAOYSA-N
Synonym 2-4-methyl-1,3-thiazol-2-yl acetonitrile, 2-4-methylthiazol-2-yl acetonitrile, 4-methyl-1,3-thiazol-2-yl acetonitrile, 4-methyl-1,3-thiazole-2-yl acetonitrile, 2-thiazoleacetonitrile,4-methyl, 2-4-methyl-1,3-thiazol-2-yl ethanenitrile, 2-thiazoleacetonitrile, 4-methyl, 4-methyl-2-thiazoleacetonitrile, 4-methylthiazole-2-acetonitrile, 4-methylthiazol-2-yl-acetonitrile
PubChem CID 737184
IUPAC Name 2-(4-methyl-1,3-thiazol-2-yl)acetonitrile
SMILES CC1=CSC(CC#N)=N1

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Quantity 10 g
Formula Weight 138.19
Percent Purity 97%
Chemical Name or Material 2-(4-Methyl-1, 3-thiazol-2-yl)acetonitrile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.