missing translation for 'onlineSavingsMsg'
Learn More

2-(4-Methoxyphenyl)malondialdehyde, 95%

Product Code. 10627771 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
2.5 g
Packaging:
Glass bottle
Unit Size:
2.5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10627771 2.5 g Glass bottle 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10627771 Supplier Thermo Scientific Acros Supplier No. 425230025
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 65192-28-1 | C10H10O3 | 178.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 65192-28-1
CAS Min % 94.0
CAS Max % 100.0
Melting Point 145°C to 150°C
Color Beige-Yellow to White
Infrared Spectrum Authentic
Assay Percent Range 95%
Packaging Glass bottle
Molecular Formula C10H10O3
MDL Number MFCD00216504
Quantity 2.5 g
Synonym 2-4-methoxyphenyl malondialdehyde, 2-4-methoxyphenyl malonaldehyde, 2-4-methoxyphenyl propanedial, 4-methoxyphenyl malonaldehyde, 2-p-methoxyphenyl malonaldehyde, propanedial, 4-methoxyphenyl, 2-4-methoxyphenyl propane-1,3-dial
Solubility Information Solubility in water: soluble. Other solubilities: soluble in alkali
InChI Key SXPCHOYAUFFORX-UHFFFAOYSA-N
SMILES COC1=CC=C(C=C1)C(C=O)C=O
IUPAC Name 2-(4-methoxyphenyl)propanedial
Molecular Weight (g/mol) 178.19
PubChem CID 3849370
Formula Weight 178.19
Percent Purity 95%
Physical Form Powder
Chemical Name or Material 2-(4-Methoxyphenyl)malondialdehyde, 95%
Show More Show Less
compliance-icons
Product Identifier
  • 2-(4-Methoxyphenyl)malondialdehyde
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.