missing translation for 'onlineSavingsMsg'
Learn More

2,4-Lutidine, 98+%

Product Code. 10497510 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
100 mL
500 mL
Packaging:
Glass bottle
Unit Size:
100mL
500mL
This item is not returnable. View return policy

Product Code. 10497510

Brand: Thermo Scientific Acros 125185000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 108-47-4 | C7H9N | 107.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108-47-4
Molecular Formula C7H9N
Molecular Weight (g/mol) 107.16
MDL Number MFCD00006337
InChI Key JYYNAJVZFGKDEQ-UHFFFAOYSA-N
Synonym 2,4-lutidine, pyridine, 2,4-dimethyl, alpha,gamma-dimethylpyridine, 2,4-lutidene, unii-83903uj0ww, 2,4-dimethyl-pyridine, .alpha.,.gamma.-dimethylpyridine, 2,4-dimethyl pyridine, 2.4-lutidine, 2,4 lutidine
PubChem CID 7936
IUPAC Name 2,4-dimethylpyridine
SMILES CC1=CC=NC(C)=C1

Specifications

CAS Min % 98.0
CAS Max % 100.0
Melting Point -60.0°C
Density 0.9200g/mL
Boiling Point 159.0°C
Flash Point 47°C
Assay Percent Range 98% min. (GC)
Packaging Glass bottle
Refractive Index 1.4990 to 1.5010
Quantity 500 mL
Beilstein , 244
Fieser 06,224
Specific Gravity 0.92
Solubility Information Solubility in water: 15g/100 g (20°C). Other solubilities: soluble in alcohol,ether,acetone and,benzene
Formula Weight 107.15
Percent Purity 98+%
Chemical Name or Material 2, 4-Lutidine, 98+%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2,4-Lutidine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H301-Toxic if swallowed.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.