missing translation for 'onlineSavingsMsg'
Learn More

2,4-Lutidine, 98+%

Product Code. 11459967
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11459967 100 g 100g
11449967 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11459967 Supplier Thermo Scientific Alfa Aesar Supplier No. B22913.22
 Restricted Item
Estimated Shipment: 28-09-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 108-47-4 | C7H9N | 107.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108-47-4
Molecular Formula C7H9N
Molecular Weight (g/mol) 107.16
MDL Number MFCD00006337
InChI Key JYYNAJVZFGKDEQ-UHFFFAOYSA-N
Synonym 2,4-lutidine, pyridine, 2,4-dimethyl, alpha,gamma-dimethylpyridine, 2,4-lutidene, unii-83903uj0ww, 2,4-dimethyl-pyridine, .alpha.,.gamma.-dimethylpyridine, 2,4-dimethyl pyridine, 2.4-lutidine, 2,4 lutidine
PubChem CID 7936
IUPAC Name 2,4-dimethylpyridine
SMILES CC1=CC=NC(C)=C1

Specifications

Melting Point -60°C
Density 0.931
Boiling Point 158°C to 160°C
Flash Point 47°C (117°F)
Odor Unpleasant
Refractive Index 1.5
Quantity 100 g
UN Number UN1992
Formula Weight 107.16
Percent Purity ≥98%
Chemical Name or Material 2,4-Lutidine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.