missing translation for 'onlineSavingsMsg'
Learn More

2-(4-Hydroxyphenylazo)benzoic acid, 97%

Product Code. 11404113
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11404113 50 g 50g
11494103 10 g 10g
11414113 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11404113 Supplier Thermo Scientific Alfa Aesar Supplier No. A10333.18
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1634-82-8 | C13H10N2O3 | 242.234 g/mol

Indicator used in determination of serum albumin

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1634-82-8
Molecular Formula C13H10N2O3
Molecular Weight (g/mol) 242.234
MDL Number MFCD00002428
InChI Key FBVSMDPNVYJNON-UHFFFAOYSA-N
Synonym 2-4-hydroxyphenylazo benzoic acid, haba, 2-p-hydroxyphenylazo benzoic acid, benzoic acid, 2-4-hydroxyphenyl azo, o-p-hydroxyphenylazo benzoic acid, 2-4'-hydroxyphenyl-azo benzoic acid, hbaba, 2-4-hydroxyphenyl diazenyl benzoic acid, 2-2-4-hydroxyphenyl diazen-1-yl benzoic acid, benzoic acid, o-p-hydroxyphenyl azo
PubChem CID 5357439
IUPAC Name 2-[2-(4-oxocyclohexa-2,5-dien-1-ylidene)hydrazinyl]benzoic acid
SMILES C1=CC=C(C(=C1)C(=O)O)NN=C2C=CC(=O)C=C2

Specifications

Melting Point 205°C to 207°C
Quantity 50 g
Beilstein 1842696
Formula Weight 242.23
Percent Purity ≥98%
Chemical Name or Material 2- (4-Hydroxyphenylazo)benzoic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.