missing translation for 'onlineSavingsMsg'
Learn More

2-(4-Hydroxyphenyl)propionic acid, 98%

Product Code. 11431457
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11431457 50 g 50g
11421457 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11431457 Supplier Thermo Scientific Alfa Aesar Supplier No. B24938.18
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 938-96-5 | C9H10O3 | 166.176 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 938-96-5
Molecular Formula C9H10O3
Molecular Weight (g/mol) 166.176
MDL Number MFCD00128847
InChI Key ZHMMPVANGNPCBW-UHFFFAOYSA-N
Synonym 2-4-hydroxyphenyl propanoic acid, 2-4-hydroxyphenyl propionic acid, 4-hydroxyhydratropate, 4-hydroxyhydratropic acid, 4-hydroxyphenyl-2-propionic acid, 2-4'-hydroxyphenyl-propionic acid, 2-4-hydroxyphenyl propionate, acmc-209ro0, 4-hydroxyphenyl propanoic acid, methyl 4-hydroxyphenylacetic acid
PubChem CID 102526
ChEBI CHEBI:1868
IUPAC Name 2-(4-hydroxyphenyl)propanoic acid
SMILES CC(C1=CC=C(C=C1)O)C(=O)O

Specifications

Melting Point 130°C to 132°C
Quantity 50 g
Formula Weight 166.18
Percent Purity 98%
Chemical Name or Material 2-(4-Hydroxyphenyl)propionic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.