missing translation for 'onlineSavingsMsg'
Learn More

2-(4-Hydroxyphenyl)-D-glycine methyl ester hydrochloride, 95%, Thermo Scientific™

Product Code. 15422579
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 15422579

Brand: Thermo Scientific Alfa Aesar H65601.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 57591-61-4
Molecular Formula C9H12ClNO3
Molecular Weight (g/mol) 217.65
MDL Number MFCD01862152
InChI Key UYCKVJUNDXPDJH-UHFFFAOYNA-N
Synonym r-amino-4-hydroxyphenyl acetic acid methyl ester hydrochloride, r-methyl 2-amino-2-4-hydroxyphenyl acetate hydrochloride, methyl 2r-2-amino-2-4-hydroxyphenyl acetate hydrochloride, d-4-hydroxyphenylglycine methyl ester hydrochloride, r-methyl 2-amino-2-4-hydroxyphenyl acetate hcl, d-p-hydroxyphenylglycine methyl ester hydrochloride, ksc495s1b, methyl r-amino-4-hydroxyphenyl acetate hydrochloride, methyl 2r-amino 4-hydroxyphenyl acetate-hydrogen chloride 1/1
PubChem CID 13215089
IUPAC Name methyl 2-amino-2-(4-hydroxyphenyl)acetate hydrochloride
SMILES Cl.COC(=O)C(N)C1=CC=C(O)C=C1

Specifications

Melting Point 187°C to 190°C
Quantity 5 g
Formula Weight 217.65
Percent Purity 95%
Physical Form Solid
Chemical Name or Material 2-(4-Hydroxyphenyl)-D-glycine methyl ester hydrochloride
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.