missing translation for 'onlineSavingsMsg'
Learn More

2-(4-Hydroxyphenyl)acetamide, 97%, Maybridge™

Product Code. 10245724
Change view
Click to view available options
Quantity:
50 g
100 g
Packaging:
Amber Glass Bottle
Unit Size:
100g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10245724 50 g Amber Glass Bottle 50g
10366944 100 g Amber Glass Bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10245724 Supplier Thermo Scientific Maybridge Supplier No. SB01670EE
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 17194-82-0
Molecular Formula C8H9NO2
Molecular Weight (g/mol) 151.165
InChI Key YBPAYPRLUDCSEY-UHFFFAOYSA-N
Synonym 4-hydroxyphenylacetamide, 2-4-hydroxyphenyl acetamide, p-hydroxyphenylacetamide, 4-hydroxybenzeneacetamide, benzeneacetamide, 4-hydroxy, p-carbamoylmethyl phenol, p-hydroxyphenyl acetamide, 2-p-hydroxyphenyl acetamide, 4-hydroxyphenyl acetamide, unii-0hy0n4itn4
PubChem CID 86986
IUPAC Name 2-(4-hydroxyphenyl)acetamide
SMILES C1=CC(=CC=C1CC(=O)N)O

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 50 g
Formula Weight 151.16
Percent Purity 97%
Chemical Name or Material 2-(4-Hydroxyphenyl)acetamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.