missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-(4-Hydroxyphenoxy-2,3,5,6-d4)-propanoic Acid, TRC
Specifications
Specifications
| Chemical Name or Material | 2-(4-Hydroxyphenoxy)propanoic Acid D4 (phenoxy-2,3,5,6-D4) |
| CAS | 127893-32-7 |
| Molecular Formula | C9 2H4 H6 O4 |
| Quantity | 5 mg |
| Synonym | Propanoic acid, 2-(4-hydroxyphenoxy-2,3,5,6-d4)- (9CI), 2-(4-Hydroxyphenoxy)propanoic acid D4 (phenoxy-2,3,5,6-D4), 2-(4-Hydroxyphenoxy-2,3,5,6-d4)propanoic acid, (±)-2-(4-Hydroxyphenoxy-2,3,5,6-d4)propionic acid, O-(4-Hydroxyphenyl-d4)lactic acid, α-(p-Hydroxyphenoxy-d4)propionic acid |
| InChI Formula | InChI=1S/C9H10O4/c1-6(9(11)12)13-8-4-2-7(10)3-5-8/h2-6,10H,1H3,(H,11,12)/i2D,3D,4D,5D |
| SMILES | [2H]c1c([2H])c(OC(C)C(=O)O)c([2H])c([2H])c1O |
| IUPAC Name | 2-(2,3,5,6-tetradeuterio-4-hydroxyphenoxy)propanoic acid |
| Molecular Weight (g/mol) | 186.198 |
| Formula Weight | 186.083 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?