missing translation for 'onlineSavingsMsg'
Learn More

2-(4-Hydroxycyclohexyl)acetic Acid, TRC

Product Code. 30394352
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30394352 100 mg
30395948 50 mg
30391283 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30394352 Supplier Toronto Research Chemicals Supplier No. TRCH955215100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 99799-09-4
Molecular Formula C8H14O3
Molecular Weight (g/mol) 158.2
Synonym 4-Hydroxycyclohexaneacetic Acid, (4-Hydroxy-cyclohexyl)-acetic Acid
IUPAC Name 2-(4-hydroxycyclohexyl)acetic acid
SMILES OC1CCC(CC(=O)O)CC1

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C8H14O3/c9-7-3-1-6(2-4-7)5-8(10)11/h6-7,9H,1-5H2,(H,10,11)
Formula Weight 158.0943
Chemical Name or Material 2-(4-Hydroxycyclohexyl)acetic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.