missing translation for 'onlineSavingsMsg'
Learn More

2-(4-Hydroxybenzoyl)benzoic acid, TRC

Product Code. 30388823
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30388823 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30388823 Supplier Toronto Research Chemicals Supplier No. TRCH949638250MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-(4-Hydroxybenzoyl)benzoic Acid
CAS 85-57-4
Molecular Formula C14 H10 O4
Quantity 250 mg
Synonym 2-(4-Hydroxybenzoyl) Benzoic Acid
InChI Formula InChI=1S/C14H10O4/c15-10-7-5-9(6-8-10)13(16)11-3-1-2-4-12(11)14(17)18/h1-8,15H,(H,17,18)
SMILES OC(=O)c1ccccc1C(=O)c2ccc(O)cc2
IUPAC Name 2-(4-hydroxybenzoyl)benzoic acid
Molecular Weight (g/mol) 242.227
Formula Weight 242.058
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.