missing translation for 'onlineSavingsMsg'
Learn More

2,4-Hexadienoic acid, 99%

Product Code. 10172770 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
250 g
500 g
1 kg
Packaging:
Plastic bottle
Unit Size:
100g
1kg
250g
500g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10172770 500 g Plastic bottle 500g
10172720 100 g Plastic bottle 100g
10599230 250 g Plastic bottle 250g
10714621 1 kg Plastic bottle 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10172770 Supplier Thermo Scientific Acros Supplier No. 120535000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 110-44-1 | C6H8O2 | 112.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 110-44-1
Molecular Formula C6H8O2
Molecular Weight (g/mol) 112.13
MDL Number MFCD00002703
InChI Key WSWCOQWTEOXDQX-MQQKCMAXSA-N
Synonym sorbic acid, 2,4-hexadienoic acid, 2e,4e-hexa-2,4-dienoic acid, 2e,4e-hexadienoic acid, panosorb, 2-propenylacrylic acid, trans,trans-sorbic acid, sorbistat, hexadienoic acid, e,e-2,4-hexadienoic acid
PubChem CID 643460
ChEBI CHEBI:38358
IUPAC Name (2E,4E)-hexa-2,4-dienoic acid
SMILES C\C=C\C=C\C(O)=O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 132°C to 135°C
Color Cream-White or White
Flash Point 127°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Plastic bottle
Linear Formula CH3CH=CHCH=CHCO2H
Quantity 500 g
Beilstein 02, 483
Fieser 15,296
Merck Index 15, 8847
Solubility Information Solubility in water: 1.6g/L (20°C). Other solubilities: soluble in ethanol,chloroform,very soluble in diethyl ether
Formula Weight 112.13
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material 2, 4-Hexadienoic acid, 99%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.