Learn More
2-[(4-Fluorophenyl)thio]acetic acid, 97%, Thermo Scientific™
£8.15 - £144.00
Chemical Identifiers
CAS | 332-51-4 |
---|---|
Molecular Formula | C8H7FO2S |
Molecular Weight (g/mol) | 186.2 |
MDL Number | MFCD00219825 |
InChI Key | HQEROVXUKINLPI-UHFFFAOYSA-N |
Synonym | 4-fluorophenylthio acetic acid, 2-4-fluorophenylthio acetic acid, 2-4-fluorophenyl thio acetic acid, 4-fluorophenyl thio acetic acid, 4-fluorophenyl sulfanyl acetic acid, maybridge1_004918, acmc-20apn5, 4-fluoro-phenylsulfanyl-acetic acid, 2-4-fluorophenyl sulfanylacetic acid |
PubChem CID | 736197 |
IUPAC Name | 2-(4-fluorophenyl)sulfanylacetic acid |
SMILES | C1=CC(=CC=C1F)SCC(=O)O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10561302
|
Thermo Scientific Maybridge
KM09206DA |
1 g |
£8.15
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10111272
|
Thermo Scientific Maybridge
KM09206EA |
10 g |
£35.75
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10001484
|
Thermo Scientific Maybridge
KM09206EE |
50 g |
£144.00
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Chemical Identifiers
332-51-4 | |
186.2 | |
HQEROVXUKINLPI-UHFFFAOYSA-N | |
736197 | |
C1=CC(=CC=C1F)SCC(=O)O |
C8H7FO2S | |
MFCD00219825 | |
4-fluorophenylthio acetic acid, 2-4-fluorophenylthio acetic acid, 2-4-fluorophenyl thio acetic acid, 4-fluorophenyl thio acetic acid, 4-fluorophenyl sulfanyl acetic acid, maybridge1_004918, acmc-20apn5, 4-fluoro-phenylsulfanyl-acetic acid, 2-4-fluorophenyl sulfanylacetic acid | |
2-(4-fluorophenyl)sulfanylacetic acid |
Specifications
332-51-4 | |
97% | |
C8H7FO2S | |
MFCD00219825 | |
HQEROVXUKINLPI-UHFFFAOYSA-N | |
2-(4-fluorophenyl)sulfanylacetic acid | |
736197 | |
97% |
97.0 | |
Amber Glass Bottle | |
FC6H4SCH2CO2H | |
4-fluorophenylthio acetic acid, 2-4-fluorophenylthio acetic acid, 2-4-fluorophenyl thio acetic acid, 4-fluorophenyl thio acetic acid, 4-fluorophenyl sulfanyl acetic acid, maybridge1_004918, acmc-20apn5, 4-fluoro-phenylsulfanyl-acetic acid, 2-4-fluorophenyl sulfanylacetic acid | |
C1=CC(=CC=C1F)SCC(=O)O | |
186.2 | |
186.21 | |
2-[(4-Fluorophenyl)thio]acetic acid |
Safety and Handling
GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.