missing translation for 'onlineSavingsMsg'
Learn More

2-(4-Fluorophenyl)-5-[(2-methylphenyl)methyl]thiophene, TRC

Product Code. 30401411
Change view
Click to view available options
Quantity:
50 mg
250 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30401411 50 mg
30395131 250 mg
30408260 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30401411 Supplier Toronto Research Chemicals Supplier No. TRCF38367050MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2005454-69-1
Molecular Formula C18H15FS
Molecular Weight (g/mol) 282.38
Synonym 2-(4-Fluorophenyl)-5-(2-methylbenzyl)thiophene
IUPAC Name 2-(4-fluorophenyl)-5-[(2-methylphenyl)methyl]thiophene
SMILES Cc1ccccc1Cc2ccc(s2)c3ccc(F)cc3

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C18H15FS/c1-13-4-2-3-5-15(13)12-17-10-11-18(20-17)14-6-8-16(19)9-7-14/h2-11H,12H2,1H3
Formula Weight 282.0878
Chemical Name or Material 2-(4-Fluorophenyl)-5-[(2-methylphenyl)methyl]thiophene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.