missing translation for 'onlineSavingsMsg'
Learn More

2-(4-Fluorophenoxy)nicotinic acid, 97+%, Thermo Scientific™

Product Code. 11457104
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11457104 1 g 1g
11467104 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11457104 Supplier Thermo Scientific Alfa Aesar Supplier No. B21533.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 54629-13-9
Molecular Formula C12H8FNO3
Molecular Weight (g/mol) 233.198
MDL Number MFCD00833409
InChI Key SDZUYDOXBXHDCE-UHFFFAOYSA-N
Synonym 2-4-fluorophenoxy nicotinic acid, 2-4-fluorophenoxy pyridine-3-carboxylic acid, 2-4-fluoro-phenoxy-nicotinic acid, pubchem16292, acmc-1akqk, maybridge1_000048, 2-4-fluorophenoxy-3-pyridinecarboxylic acid, 3-pyridinecarboxylicacid, 2-4-fluorophenoxy, 3-pyridinecarboxylic acid, 2-4-fluorophenoxy
PubChem CID 605123
IUPAC Name 2-(4-fluorophenoxy)pyridine-3-carboxylic acid
SMILES C1=CC(=C(N=C1)OC2=CC=C(C=C2)F)C(=O)O

Specifications

Melting Point 182°C to 185°C
Quantity 1 g
Formula Weight 233.2
Percent Purity ≥97%
Chemical Name or Material 2-(4-Fluorophenoxy)nicotinic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.