missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-(4-Fluorophenoxy)-N-methylbenzylamine, 96%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
2-(4-Fluorophenoxy)-N-methylbenzylamine is used as pharmaceutical-intermediates.
Notes
Air Sensitive. Store in a cool, dry condition in a well sealed container. Incompatible with oxidizing agents and air. Store under dry inert gas.
2-(4-Fluorophenoxy)-N-methylbenzylamine is used as pharmaceutical-intermediates.
Notes
Air Sensitive. Store in a cool, dry condition in a well sealed container. Incompatible with oxidizing agents and air. Store under dry inert gas.
Specifications
Spezifikation
Quantity | 250 mg |
Sensitivity | Air Sensitive |
Formula Weight | 231.27 |
Percent Purity | 96% |
Chemical Name or Material | 2-(4-Fluorophenoxy)-N-methylbenzylamine |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'