missing translation for 'onlineSavingsMsg'
Learn More

2-[4-(Ethoxycarbonyl)phenyl]-6-methyl-imidazo[1,2-a]pyridine, TRC

Product Code. 30384077
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30384077 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30384077 Supplier Toronto Research Chemicals Supplier No. TRCE891615100MG
 Restricted Item
Estimated Shipment: 14-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-[4-(Ethoxycarbonyl)phenyl]-6-methyl-imidazo[1,2-a]pyridine
CAS 109461-69-0
Molecular Formula C17 H16 N2 O2
Quantity 100 mg
Synonym 4-(6-Methylimidazo[1,2-a]pyridin-2-yl)benzoic Acid Ethyl Ester
InChI Formula InChI=1S/C17H16N2O2/c1-3-21-17(20)14-7-5-13(6-8-14)15-11-19-10-12(2)4-9-16(19)18-15/h4-11H,3H2,1-2H3
SMILES CCOC(=O)c1ccc(cc1)c2cn3cc(C)ccc3n2
IUPAC Name ethyl 4-(6-methylimidazo[1,2-a]pyridin-2-yl)benzoate
Molecular Weight (g/mol) 280.32
Formula Weight 280.1212
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.