missing translation for 'onlineSavingsMsg'
Learn More

2,4-Diphenyl-1,3-thiazole-5-sulfonyl chloride, ≥97%, Thermo Scientific™

Product Code. 10297822
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10297822 1 g Amber Glass Bottle 1g
10557304 250 mg Amber Glass Bottle 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10297822 Supplier Thermo Scientific Maybridge Supplier No. CC42303DA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 868755-57-1
Molecular Formula C15H10ClNO2S2
Molecular Weight (g/mol) 335.82
MDL Number MFCD08271911
InChI Key AWNJOBYLGNWNCQ-UHFFFAOYSA-N
Synonym diphenyl-1,3-thiazole-5-sulfonyl chloride, 2,4-diphenylthiazole-5-sulfonyl chloride, 5-thiazolesulfonylchloride, 2,4-diphenyl, 5-chlorosulphonyl-2,4-diphenyl-1,3-thiazole
PubChem CID 18525757
SMILES ClS(=O)(=O)C1=C(N=C(S1)C1=CC=CC=C1)C1=CC=CC=C1

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
IUPAC Name 2,4-diphenyl-1,3-thiazole-5-sulfonyl chloride
Formula Weight 335.83
Percent Purity 97%
Chemical Name or Material 2, 4-Diphenyl-1, 3-thiazole-5-sulfonyl chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.