missing translation for 'onlineSavingsMsg'
Learn More

2,4-Diphenyl-1,3-thiazole-5-carboxylic acid, ≥97%, Thermo Scientific™

Product Code. 10158182
Change view
Click to view available options
Quantity:
250 mg
1 g
10 g
Unit Size:
10g
1g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10158182 250 mg 250mg
10789573 1 g 1g
10163724 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10158182 Supplier Thermo Scientific Maybridge Supplier No. CC42301CB
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 502935-47-9
Molecular Formula C16H11NO2S
Molecular Weight (g/mol) 281.329
MDL Number MFCD07348756
InChI Key KMOCHRNIGWCEJV-UHFFFAOYSA-N
Synonym 2,4-diphenylthiazole-5-carboxylic acid, diphenyl-1,3-thiazole-5-carboxylic acid, 5-thiazolecarboxylicacid, 2,4-diphenyl
PubChem CID 7131193
IUPAC Name 2,4-diphenyl-1,3-thiazole-5-carboxylic acid
SMILES C1=CC=C(C=C1)C2=C(SC(=N2)C3=CC=CC=C3)C(=O)O

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 250 mg
Formula Weight 281.33
Percent Purity 97%
Chemical Name or Material 2, 4-Diphenyl-1, 3-thiazole-5-carboxylic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.