missing translation for 'onlineSavingsMsg'
Learn More

(2,4-Diphenyl-1,3-thiazol-5-yl)methylamine hydrochloride, ≥95%, Thermo Scientific™

Product Code. 10767285
Change view
Click to view available options
Amber Glass Bottle; 1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10767285 Amber Glass Bottle; 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10767285 Supplier Thermo Scientific Maybridge Supplier No. CC42363DA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

CAS 879896-44-3
CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Molecular Formula C16H15ClN2S
MDL Number MFCD08435866
Quantity 1 g
Synonym 2,4-diphenyl-1,3-thiazol-5-yl methylamine hydrochloride, diphenyl-1,3-thiazol-5-yl methanamine hydrochloride, 1-2,4-diphenyl-1,3-thiazol-5-yl methanamine hydrochloride, c16h14n2s.hcl, 2,4-diphenylthiazol-5-yl methanamine hydrochloride, 1-2,4-diphenyl-1,3-thiazol-5-yl methanamine-hydrogen chloride 1/1
InChI Key REDARXMRPRJWQP-UHFFFAOYSA-N
SMILES Cl.NCC1=C(N=C(S1)C1=CC=CC=C1)C1=CC=CC=C1
IUPAC Name (2,4-diphenyl-1,3-thiazol-5-yl)methanamine;hydrochloride
Molecular Weight (g/mol) 302.82
PubChem CID 18525760
Formula Weight 302.83
Percent Purity 95%
Chemical Name or Material (2, 4-Diphenyl-1, 3-thiazol-5-yl)methylamine hydrochloride
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.