missing translation for 'onlineSavingsMsg'
Learn More

(2,4-Diphenyl-1,3-thiazol-5-yl)methanol, ≥97%, Thermo Scientific™

Product Code. 10391333
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10391333 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10391333 Supplier Thermo Scientific Maybridge Supplier No. CC42309DA
 Restricted Item
Estimated Shipment: 03-02-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 864068-86-0
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Molecular Formula C16H13NOS
MDL Number MFCD08060522
Quantity 1 g
Synonym 2,4-diphenyl-1,3-thiazol-5-yl methanol, diphenyl-1,3-thiazol-5-yl methanol, 5-thiazolemethanol,2,4-diphenyl, 2,4-diphenylthiazol-5-yl methanol
InChI Key IVIJHUHOSCRMNY-UHFFFAOYSA-N
SMILES OCC1=C(N=C(S1)C1=CC=CC=C1)C1=CC=CC=C1
IUPAC Name (2,4-diphenyl-1,3-thiazol-5-yl)methanol
Molecular Weight (g/mol) 267.35
PubChem CID 7537555
Formula Weight 267.35
Percent Purity 97%
Chemical Name or Material (2, 4-Diphenyl-1, 3-thiazol-5-yl)methanol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.