missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dinitroaniline, TRC

Product Code. 30180347
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30180347 5 g 5g
30180256 25 g 25g
30179862 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30180347 Supplier Toronto Research Chemicals Supplier No. TRCD4739225G

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.
, 02 950 59 478

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 97-02-9
Molecular Formula C6 H5 N3 O4
Molecular Weight (g/mol) 183.12
Synonym Benzenamine, 2,4-dinitro- (9CI, ACI), 2,4-Dinitrobenzenamine (ACI), Aniline, 2,4-dinitro- (8CI), 1-Amino-2,4-dinitrobenzene, 2,4-Dinitroaniline, Dinitroaniline, NSC 8731
IUPAC Name 2,4-dinitroaniline
SMILES Nc1ccc(cc1[N+](=O)[O-])[N+](=O)[O-]

Specifications

Quantity 5 g
InChI Formula InChI=1S/C6H5N3O4/c7-5-2-1-4(8(10)11)3-6(5)9(12)13/h1-3H,7H2
Formula Weight 183.028
Chemical Name or Material 2,4-Dinitroaniline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.