missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dinitro-6-(1-methylheptyl)phenol, TRC

Product Code. 30134790
Change view
Click to view available options
Quantity:
1 mg
2.5 mg
25 mg
Unit Size:
1mg
2.5mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30134790 25 mg 25mg
30133322 1 mg 1mg
30010897 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30134790 Supplier Toronto Research Chemicals Supplier No. TRCD49238825MG
 Restricted Item
Estimated Shipment: 18-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3687-22-7
Molecular Formula C14 H20 N2 O5
Molecular Weight (g/mol) 296.32
Synonym 2-(1-Methylheptyl)-4,6-dinitrophenol, 2,4-DNOP, 2,4-Dinitro-6-(1-methylheptyl)phenol, 2,4-Dinitro-6-(1'-methylheptyl)phenol, 2-(2-Hydroxy-3,5-dinitrophenyl)octane, DNOP, Meptyldinocap-phenol
IUPAC Name 2,4-dinitro-6-octan-2-ylphenol
SMILES CCCCCCC(C)c1cc(cc(c1O)[N+](=O)[O-])[N+](=O)[O-]

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C14H20N2O5/c1-3-4-5-6-7-10(2)12-8-11(15(18)19)9-13(14(12)17)16(20)21/h8-10,17H,3-7H2,1-2H3
Formula Weight 296.1372
Chemical Name or Material Meptyldinocap-phenol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.