missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dinitro-1H-imidazole, TRC

Product Code. 30381190
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
Unit Size:
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30381190 500 mg 500mg
30369267 1 g -
30383346 100 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30381190 Supplier Toronto Research Chemicals Supplier No. TRCD479900500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5213-49-0
Molecular Formula C3H2N4O4
Molecular Weight (g/mol) 158.07
Synonym 2,4(5)-Dinitroimidazole, 2,4-Dinitro-1H-imidazole, 2,4-Dinitroimidazole, 2,5-Dinitroimidazole, KA 121, NSC 342704
IUPAC Name 2,5-dinitro-1H-imidazole
SMILES [O-][N+](=O)c1cnc([nH]1)[N+](=O)[O-]

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C3H2N4O4/c8-6(9)2-1-4-3(5-2)7(10)11/h1H,(H,4,5)
Formula Weight 158.0076
Chemical Name or Material 2,4-Dinitro-1H-imidazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.