missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dinitro-1-(trifluoromethoxy)benzene, 98%, Thermo Scientific™

Product Code. 11410447
Change view
Click to view available options
Quantity:
2.5 g
10 g
Unit Size:
10g
2.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11410447 10 g 10g
11400447 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11410447 Supplier Thermo Scientific Alfa Aesar Supplier No. B21790.09
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Not miscible or difficult to mix with water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 655-07-2
Molecular Formula C7H3F3N2O5
Molecular Weight (g/mol) 252.11
MDL Number MFCD00042268
InChI Key KYUUZCUXYXNPFO-UHFFFAOYSA-N
Synonym 2,4-dinitro-1-trifluoromethoxy benzene, 2,4-dinitro trifluoromethoxy benzene, 2,4-dinitrotrifluoroanisole, 2,4-dinitrotrifluoromethoxybenzene, 2,4-dinitro-1-trifluoromethoxy-benzene, acmc-20aoc3, 2,4-dinitro trifluoroanisole, a,a,a-trifluoro-2,4-dinitro-anisole, 1-trifluoromethoxy-2,4-dinitrobenzene, benzene,2,4-dinitro-1-trifluoromethoxy
PubChem CID 2737131
IUPAC Name 2,4-dinitro-1-(trifluoromethoxy)benzene
SMILES [O-][N+](=O)C1=CC=C(OC(F)(F)F)C(=C1)[N+]([O-])=O

Specifications

Melting Point -21°C
Density 1.623
Boiling Point 273°C to 274°C
Flash Point 154°C (309°F)
Quantity 10 g
UN Number UN2927
Solubility Information Not miscible or difficult to mix with water.
Formula Weight 252.11
Percent Purity 98%
Chemical Name or Material 2,4-Dinitro-1-(trifluoromethoxy)benzene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.