missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dimethylstyrene, 97%, stabilized

Product Code. 10499052 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10499052 5 g Glass bottle 5g
15234694 25 g Glass bottle 25g
15244694 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10499052 Supplier Thermo Scientific Acros Supplier No. 271150050
 Restricted Item
Estimated Shipment: 05-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 2234-20-0 | C10H12 | 132.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Kemiska identifierare

CAS 2234-20-0
Molecular Formula C10H12
Molecular Weight (g/mol) 132.21
MDL Number MFCD00014937
InChI Key OEVVKKAVYQFQNV-UHFFFAOYSA-N
Synonym 2,4-dimethylstyrene, 4-vinyl-m-xylene, styrene, 2,4-dimethyl, 1,3-dimethyl-4-vinylbenzene, 1-vinyl-2,4-dimethylbenzene, 1,3-dimethyl-4-ethenylbenzene, benzene, 1-ethenyl-2,4-dimethyl, unii-543es1o25e, styrene,4-dimethyl, acmc-1cb1s
PubChem CID 16694
IUPAC Name 1-ethenyl-2,4-dimethylbenzene
SMILES CC1=CC(=C(C=C1)C=C)C

Specifikationer

CAS Min % 96.0
CAS Max % 100.0
Melting Point -64.0°C
Density 0.9060g/mL
Boiling Point 73.0°C to 74.0°C (10.0 mmHg)
Flash Point 60°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula (CH3)2C6H3CH=CH2
Refractive Index 1.5410 to 1.5440
Quantity 5 g
Specific Gravity 0.906
Stabilizer 0.05 % (ca.) t-butylcatechol
Formula Weight 132.21
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 2, 4-Dimethylstyrene, 97%
Visa mer Visa mindre
compliance-icons compliance-icons
Produktbeteckning
  • 2,4-Dimethylstyrene, stabilized
Signalord
  • Warning
Riskkategorier
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Faroangivelser
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H226-Flammable liquid and vapour.
Skyddsangivelse
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.