missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dimethylquinoline, TRC

Product Code. 30386486
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30386486 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30386486 Supplier Toronto Research Chemicals Supplier No. TRCD480538500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2,4-Dimethylquinoline
CAS 1198-37-4
Molecular Formula C11 H11 N
Quantity 500 mg
Synonym 2,4-Dimethylquinoline, 4-Methylquinaldine, NSC 62132
InChI Formula InChI=1S/C11H11N/c1-8-7-9(2)12-11-6-4-3-5-10(8)11/h3-7H,1-2H3
SMILES Cc1cc(C)c2ccccc2n1
IUPAC Name 2,4-dimethylquinoline
Molecular Weight (g/mol) 157.21
Formula Weight 157.0891
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.