missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dimethylquinoline, 95%

Product Code. 11496750
Change view
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11496750 500 mg 500mg
11486750 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11496750 Supplier Thermo Scientific Alfa Aesar Supplier No. L08348.ME
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1198-37-4 | C11H11N | 157.216 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1198-37-4
Molecular Formula C11H11N
Molecular Weight (g/mol) 157.216
MDL Number MFCD00006760
InChI Key ZTNANFDSJRRZRJ-UHFFFAOYSA-N
PubChem CID 14536
IUPAC Name 2,4-dimethylquinoline
SMILES CC1=CC(=NC2=CC=CC=C12)C

Specifications

Density 1.06
Boiling Point 264°C to 265°C
Flash Point >110°C (230°F)
Refractive Index 1.605
Quantity 500 mg
Beilstein 114230
Synonym 4-methylquinaldine, quinoline, 2,4-dimethyl, 2,4-dimethyl-quinoline, kryptidine, methylquinaldine, pubchem5875, quinoline,4-dimethyl, 2,4-dimethyl-quinolin, quinoline,2,4-dimethyl, acmc-1bq9j
Formula Weight 157.21
Percent Purity 95%
Chemical Name or Material 2,4-Dimethylquinoline

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.