missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dimethylphenyl isocyanate, 98+%

Product Code. p-7045551
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11307576

Brand: Thermo Scientific Alfa Aesar L11126.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 51163-29-2 | C9H9NO | 147.177 g/mol

2,4-Dimethylphenyl isocyanate is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,4-Dimethylphenyl isocyanate is used as a pharmaceutical intermediate.

Solubility
Hydrolyzes in water.

Notes
Moisture sensitive. Store away from strong oxidizing agents, water/ moisture, heat, alcohols and amines.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 51163-29-2
Molecular Formula C9H9NO
Molecular Weight (g/mol) 147.177
MDL Number MFCD00013853
InChI Key QUOBVYPFBJUOAJ-UHFFFAOYSA-N
Synonym 2,4-dimethylphenyl isocyanate, 2,4-dimethylphenylisocyanate, benzene, 1-isocyanato-2,4-dimethyl, 2,4-dimethylbenzenisocyanate, acmc-20aoc8, 1-isocyanato-2,4-dimethyl-benzene
PubChem CID 5207585
IUPAC Name 1-isocyanato-2,4-dimethylbenzene
SMILES CC1=CC(=C(C=C1)N=C=O)C

Specifications

Density 1.041
Boiling Point 114°C to 115°C (2 mmHg)
Quantity 1 g
Refractive Index 1.535
UN Number UN2206
Beilstein 386756
Solubility Information Hydrolyzes in water.
Formula Weight 147.18
Percent Purity ≥98%
Chemical Name or Material 2,4-Dimethylphenyl isocyanate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.