missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dimethylpentane, 98+%

Product Code. p-7043939
Click to view available options
Quantity:
2 g
10 g
Unit Size:
10g
2g
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.

Product Code. 11362216

Brand: Thermo Scientific Alfa Aesar L03570.04

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.

CAS: 108-08-7 | C7H16 | 100.205 g/mol

It is not only used for synthesizing WeiNiGuan, but also used in binder and coating industry. It is used for synthesizing resin fiber. It is also used as intermediates and adhesives in oil drop coagulation thickener.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is not only used for synthesizing WeiNiGuan, but also used in binder and coating industry. It is used for synthesizing resin fiber. It is also used as intermediates and adhesives in oil drop coagulation thickener.

Solubility
Soluble in acetone, benzene, chloroform, alcohol and ether.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108-08-7
Molecular Formula C7H16
Molecular Weight (g/mol) 100.205
MDL Number MFCD00008945
InChI Key BZHMBWZPUJHVEE-UHFFFAOYSA-N
PubChem CID 7907
IUPAC Name 2,4-dimethylpentane
SMILES CC(C)CC(C)C

Specifications

Melting Point ∼-123°C
Boiling Point 79°C to 80°C
Flash Point −12°C (10°F)
Quantity 2 g
UN Number UN1206
Beilstein 1696855
Synonym pentane, 2,4-dimethyl, unii-4jt8q9qohi, diisopropylmethane, 4jt8q9qohi, 2,4-dimethylpentane, standard for gc, dimethylpentane, heptan und isomere, 2,4-dimethyl-pentan, pentane,2,4-dimethyl, 2,4-dimethylpentane
Formula Weight 100.21
Percent Purity ≥98%
Chemical Name or Material 2,4-Dimethylpentane

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Nom du produit

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.

Merci de nous aider à améliorer notre site web. Votre commentaire a été soumis