missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dimethylpentane, 98+%

Product Code. 11362216
Change view
Click to view available options
Quantity:
2 g
10 g
Unit Size:
10g
2g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11362216 2 g 2g
11372216 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: This product is subject to excise duty on mineral oils in accordance with European Directive 2003/96/EC.

We regret to inform you that this product is not available for the Canary Islands, Ceuta and Melilla. We apologise for the inconvenience.
Product Code. 11362216 Supplier Thermo Scientific Alfa Aesar Supplier No. L03570.04
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: This product is subject to excise duty on mineral oils in accordance with European Directive 2003/96/EC.

We regret to inform you that this product is not available for the Canary Islands, Ceuta and Melilla. We apologise for the inconvenience.

CAS: 108-08-7 | C7H16 | 100.205 g/mol

It is not only used for synthesizing WeiNiGuan, but also used in binder and coating industry. It is used for synthesizing resin fiber. It is also used as intermediates and adhesives in oil drop coagulation thickener.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is not only used for synthesizing WeiNiGuan, but also used in binder and coating industry. It is used for synthesizing resin fiber. It is also used as intermediates and adhesives in oil drop coagulation thickener.

Solubility
Soluble in acetone, benzene, chloroform, alcohol and ether.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108-08-7
Molecular Formula C7H16
Molecular Weight (g/mol) 100.205
MDL Number MFCD00008945
InChI Key BZHMBWZPUJHVEE-UHFFFAOYSA-N
PubChem CID 7907
IUPAC Name 2,4-dimethylpentane
SMILES CC(C)CC(C)C

Specifications

Melting Point ∼-123°C
Boiling Point 79°C to 80°C
Flash Point −12°C (10°F)
Quantity 2 g
UN Number UN1206
Beilstein 1696855
Synonym pentane, 2,4-dimethyl, unii-4jt8q9qohi, diisopropylmethane, 4jt8q9qohi, 2,4-dimethylpentane, standard for gc, dimethylpentane, heptan und isomere, 2,4-dimethyl-pentan, pentane,2,4-dimethyl, 2,4-dimethylpentane
Formula Weight 100.21
Percent Purity ≥98%
Chemical Name or Material 2,4-Dimethylpentane

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.