missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dimethylimidazole, 97%, Thermo Scientific Chemicals

Product Code. 11453954
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

It is employed as a curing agent of epoxy resins. It is also widely used in the bonding, coating, capsulation of epoxy resins, or used for composite materials. It is also employed as medical raw material or used in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed as a curing agent of epoxy resins. It is also widely used in the bonding, coating, capsulation of epoxy resins, or used for composite materials. It is also employed as medical raw material or used in organic synthesis.

Solubility
Soluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed. Store in cool, dry conditions in well ventilated place.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 930-62-1
Molecular Formula C5H8N2
Molecular Weight (g/mol) 96.13
MDL Number MFCD00022365
InChI Key LLPKQRMDOFYSGZ-UHFFFAOYSA-N
Synonym 2,4-dimethylimidazole, 2,4-dimethyl-1h-imidazole, 1h-imidazole, 2,4-dimethyl, 2,4-dimethyl-3h-imidazole, 2,4-dimethyl-1h-imidazol, imidazole, 2,4-dimethyl, 2,4-dimethyl, 2,5-dimethylimidazole, imidazole,4-dimethyl, pubchem18587
PubChem CID 70259
IUPAC Name 2,5-dimethyl-1H-imidazole
SMILES CC1=CN=C(C)N1

Specifications

Melting Point 84°C to 88°C
Boiling Point 266°C (733 mmHg)
Quantity 5 g
UN Number UN3263
Beilstein 1639
Solubility Information Soluble in water.
Formula Weight 96.13
Percent Purity 97%
Chemical Name or Material 2,4-Dimethylimidazole

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'