missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dimethylimidazole, 97%

Product Code. 11463954
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11463954 25 g 25g
11453954 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11463954 Supplier Thermo Scientific Alfa Aesar Supplier No. A11949.14
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 930-62-1 | C5H8N2 | 96.13 g/mol

It is employed as a curing agent of epoxy resins. It is also widely used in the bonding, coating, capsulation of epoxy resins, or used for composite materials. It is also employed as medical raw material or used in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed as a curing agent of epoxy resins. It is also widely used in the bonding, coating, capsulation of epoxy resins, or used for composite materials. It is also employed as medical raw material or used in organic synthesis.

Solubility
Soluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed. Store in cool, dry conditions in well ventilated place.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 930-62-1
Molecular Formula C5H8N2
Molecular Weight (g/mol) 96.13
MDL Number MFCD00022365
InChI Key LLPKQRMDOFYSGZ-UHFFFAOYSA-N
Synonym 2,4-dimethylimidazole, 2,4-dimethyl-1h-imidazole, 1h-imidazole, 2,4-dimethyl, 2,4-dimethyl-3h-imidazole, 2,4-dimethyl-1h-imidazol, imidazole, 2,4-dimethyl, 2,4-dimethyl, 2,5-dimethylimidazole, imidazole,4-dimethyl, pubchem18587
PubChem CID 70259
IUPAC Name 2,5-dimethyl-1H-imidazole
SMILES CC1=CN=C(C)N1

Specifications

Melting Point 84°C to 88°C
Boiling Point 266°C (733 mmHg)
Quantity 25 g
UN Number UN3263
Beilstein 1639
Solubility Information Soluble in water.
Formula Weight 96.13
Percent Purity 97%
Chemical Name or Material 2,4-Dimethylimidazole
compliance-icons
Product Identifier
  • 2,4-Dimethylimidazole
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: 2,4-Dimethyl-1H-imidazole

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.