missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dimethylimidazole, 90%, Tech.

Product Code. 10449910 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10449910 25 g Glass bottle 25g
10338730 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10449910 Supplier Thermo Scientific Acros Supplier No. 210320250
 Restricted Item
Estimated Shipment: 19-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 930-62-1 | C5H8N2 | 96.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 930-62-1
Molecular Formula C5H8N2
Molecular Weight (g/mol) 96.13
MDL Number MFCD00022365
InChI Key LLPKQRMDOFYSGZ-UHFFFAOYSA-N
Synonym 2,4-dimethylimidazole, 2,4-dimethyl-1h-imidazole, 1h-imidazole, 2,4-dimethyl, 2,4-dimethyl-3h-imidazole, 2,4-dimethyl-1h-imidazol, imidazole, 2,4-dimethyl, 2,4-dimethyl, 2,5-dimethylimidazole, imidazole,4-dimethyl, pubchem18587
PubChem CID 70259
IUPAC Name 2,5-dimethyl-1H-imidazole
SMILES CC1=CN=C(C)N1

Specifications

CAS Min % 88.0
CAS Max % 100.0
Melting Point 85.0°C to 87.0°C
Color Yellow to Orange
Boiling Point 266.0°C (733.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 88% min. (GC)
Packaging Glass bottle
Quantity 25 g
Beilstein 23, V,5, 172
Formula Weight 96.13
Percent Purity 90%
Grade Technical
Physical Form Viscous Liquid or Weak Mass
Chemical Name or Material 2, 4-Dimethylimidazole, tech., 90%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2,4-Dimethylimidazole
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.